3,5-Difluorophenol ≥99%

197572-10G 197572-1G
220042-099EA 294 USD
220042-099 220042-100
3,5-Difluorophenol ≥99%
3,5-Difluorophenol
Formula: C₆H₄F₂O
MW: 130.09 g/mol
Boiling Pt: 65 - 68 °C (15 -rr)
Melting Pt: 55 °C
Density: 1.566 g/cm³ (20 °C)
MDL Number: MFCD00002255
CAS Number: 2713-34-0
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR