Triisopropyl borate ≥98%

Synonyms: Tri-i-propylborate

197335-750G 197335-80G
220042-080EA 156.46 USD
220042-080 220042-081
Triisopropyl borate ≥98%
Triisopropyl borate
Formula: C₉H₂₁BO₃
MW: 188.07 g/mol
Boiling Pt: 139 °C (1013 hPa)
Melting Pt: –59 °C
Density: 0.82 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008872
CAS Number: 5419-55-6
EINECS: 226-529-9
UN: 2616
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR