5-(Trifluoromethyl)-m-anisidine ≥99%

Synonyms: 3-Amino-5-(trifluoromethyl)anisole, 3-Amino-5-methoxybenzotrifluoride, 3-Methoxy-5-(trifluoromethyl)aniline, 5-(Trifluoromethyl)-3-anisidine, 5-(Trifluoromethyl)-3-methoxyaniline

196932-1G 196932-5G
220042-056EA 66.23 USD
220042-056 220042-057
5-(Trifluoromethyl)-m-anisidine ≥99%
5-(Trifluoromethyl)-m-anisidine
Formula: C₈H₈F₃NO
MW: 191.15 g/mol
MDL Number: MFCD00000393
CAS Number: 349-55-3
EINECS: 206-487-8
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR