1,3,5-Trifluorobenzene ≥97%

196851-5G 196851-25G
220041-200EA 114.15 USD
220041-200 220041-199
1,3,5-Trifluorobenzene ≥97%
1,3,5-Trifluorobenzene
Formula: C₆H₃F₃
MW: 132.09 g/mol
Boiling Pt: 75 °C (1013 hPa)
Density: 1.277 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000333
CAS Number: 372-38-3
EINECS: 206-751-2
UN: 2387
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR