2,3,5,6-Tetrafluorophenol ≥97%

Warning

196789-5G 196789-25G
220041-191EA 55.14 USD
220041-191 220041-190
2,3,5,6-Tetrafluorophenol ≥97%
2,3,5,6-Tetrafluorophenol
Formula: C₆H₂F₄O
MW: 166.07 g/mol
Boiling Pt: 140 °C (1013 hPa)
Melting Pt: 30 °C
MDL Number: MFCD00002157
CAS Number: 769-39-1
EINECS: 212-209-6
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR