4-tert-Butylpyrocatechol

Danger

Synonyms: 4-(tert-Butyl)benzene-1,2-diol, 4-tert-Butylcatechol

19670-100G 19670-500G
220041-746EA 40.71 USD
220041-746 220041-747
4-tert-Butylpyrocatechol
4-tert-Butylpyrocatechol
Formula: C₁₀H₁₄O₂
MW: 166.22 g/mol
Boiling Pt: 285 °C (1013 hPa)
Melting Pt: 53 °C
Density: 1.05 g/cm³ (60 °C)
Flash Pt: 129 °C
Storage Temperature: Ambient
MDL Number: MFCD00002201
CAS Number: 98-29-3
EINECS: 202-653-9
UN: 2923
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR