1-Fluoronaphthalene

Warning

196657-25G 196657-5G
220041-738EA 123.85 USD
220041-738 220041-739
1-Fluoronaphthalene
1-Fluoronaphthalene
Formula: C₁₀H₇F
MW: 146.16 g/mol
Boiling Pt: 215 °C (1013 hPa)
Melting Pt: –12.5 °C
Density: 1.135 g/cm³ (0 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003873
CAS Number: 321-38-0
EINECS: 206-287-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR