2-Fluorobenzonitrile ≥98%

Warning

Synonyms: o-Fluorobenzonitrile

196649-25G 196649-5G
220041-736EA 49.54 USD
220041-736 220041-737
2-Fluorobenzonitrile ≥98%
2-Fluorobenzonitrile
Formula: C₇H₄FN
MW: 121.11 g/mol
Boiling Pt: 190 °C (1013 hPa)
Melting Pt: 35…37 °C
Density: 1.11 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00001773
CAS Number: 394-47-8
EINECS: 206-897-7
UN: 3276
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR