2,6-Difluoroaniline ≥97%

Danger

196614-100G 196614-5G 196614-25G
220041-558EA 144.31 USD
220041-558 220041-733 220041-559
2,6-Difluoroaniline ≥97%
2,6-Difluoroaniline
Formula: C₆H₅F₂N
MW: 129.11 g/mol
Boiling Pt: 51 °C (1013 hPa)
Density: 1.199 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007655
CAS Number: 5509-65-9
EINECS: 226-853-0
UN: 1992
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR