4-Bromothioanisole ≥97%

Warning

Synonyms: 1-Bromo-4-(methylthio)benzene, 4-Bromophenyl methyl sulphide

196525-25G 196525-100G
220041-550EA 76.68 USD
220041-550 220041-549
4-Bromothioanisole ≥97%
4-Bromothioanisole
Formula: C₇H₇BrS
MW: 203.1 g/mol
Boiling Pt: 242 °C (1013 hPa)
Melting Pt: 35 °C
Density: 1.4584 g/cm³ (39 °C)
MDL Number: MFCD00000102
CAS Number: 104-95-0
EINECS: 203-255-8
UN: 3335
ADR: 9,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR