9-(Chloromethyl)anthracene ≥98%

Warning

196517-10G 196517-2.5G
220041-547EA 92.08 USD
220041-547 220041-548
9-(Chloromethyl)anthracene ≥98%
9-(Chloromethyl)anthracene
Formula: C₁₅H₁₁Cl
MW: 226.71 g/mol
Boiling Pt: 371 °C (1013 hPa)
Melting Pt: 142 °C
Storage Temperature: Ambient
MDL Number: MFCD00001263
CAS Number: 24463-19-2
EINECS: 246-274-7
UN: 3261
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR