(4-Biphenyl)acetic acid ≥98%

Danger

Synonyms: 4-Biphenylacetic acid

196487-25G 196487-5G
220041-544EA 417.69 USD
220041-544 220041-545
(4-Biphenyl)acetic acid ≥98%
(4-Biphenyl)acetic acid
Formula: (C₆H₅C₆H₅)CH₂COOH
MW: 212.25 g/mol
Boiling Pt: 365 °C (1013 hPa)
Melting Pt: 160…163 °C
Storage Temperature: Ambient
MDL Number: MFCD00004351
CAS Number: 5728-52-9
EINECS: 227-233-2
UN: 2811
ADR: 6.1,III
Merck Index: 13,03979

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR