4-Phenylcyclohexanone ≥98%

Synonyms: 4-Phenylcyclohexan-1-one, 1-Oxo-4-phenylcyclohexane

196231-50G 196231-10G
220041-726EA 190.62 USD
220041-726 220041-725
4-Phenylcyclohexanone ≥98%
4-Phenylcyclohexanone
Formula: C₁₂H₁₄O
MW: 174.24 g/mol
Boiling Pt: 294 °C (1013 hPa)
Melting Pt: 76…78 °C
Storage Temperature: Ambient
MDL Number: MFCD00001641
CAS Number: 4894-75-1
EINECS: 225-517-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR