Ethyl isothiocyanatoformate ≥98%

Synonyms: Ethoxycarbonyl isothiocyanate

196126-25G 196126-5G
220042-043EA 269.23 USD
220042-043 220042-044
Ethyl isothiocyanatoformate ≥98%
Ethyl isothiocyanatoformate
Formula: C₄H₅NO₂S
MW: 131.16 g/mol
MDL Number: MFCD00004814
CAS Number: 16182-04-0
EINECS: 240-318-9

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR