Tetramethylammonium bromide

Danger

195758-100G 195758-500G
220042-014EA 26.77 USD
220042-014 220042-015
Tetramethylammonium bromide
Tetramethylammonium bromide
Formula: C₄H₁₂BrN
MW: 154.05 g/mol
Boiling Pt: ? 360 °C (1013 hPa)
Density: 1.56 g/cm³ (20 °C)
MDL Number: MFCD00011626
CAS Number: 64-20-0
EINECS: 200-581-2
UN: 2811
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR