cis-1,4-Dichloro-2-butene ≥95%

195707-25G 195707-100G 195707-5G
220042-005EA 95.09 USD
220042-005 220042-004 220042-006
cis-1,4-Dichloro-2-butene ≥95%
cis-1,4-Dichloro-2-butene
Formula: C₄H₆Cl₂
MW: 125 g/mol
Boiling Pt: 152 °C (1013 hPa)
Melting Pt: -48 °C
Density: 1.1363 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00062950
CAS Number: 1476-11-5
EINECS: 216-021-5
UN: 3390
ADR: 6.1,I

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR