Methoxyacetic acid ≥98%

Synonyms: 2-Methoxyacetic acid

194557-50G 194557-250G
220041-881EA 59.12 USD
220041-881 220041-880
Methoxyacetic acid ≥98%
Methoxyacetic acid
Formula: C₃H₆O₃
MW: 90.08 g/mol
Boiling Pt: 203 °C (1013 hPa)
Melting Pt: 7 °C
Density: 1.18 g/cm³ (20 °C)
MDL Number: MFCD00004308
CAS Number: 625-45-6
EINECS: 210-894-6
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR