1,3-Dibromobenzene ≥97%

Warning

194395-25G 194395-5G
220040-190EA 77 USD
220040-190 220040-191
1,3-Dibromobenzene ≥97%
1,3-Dibromobenzene
Formula: C₆H₄Br₂
MW: 235.91 g/mol
Boiling Pt: 218 °C (1013 hPa)
Melting Pt: -7 °C
Density: 1.952 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000078
CAS Number: 108-36-1
EINECS: 203-574-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR