Dimethyl isophthalate ≥99%

Synonyms: Isophthalic acid dimethyl ester

194239-500G 194239-25G 194239-3KG
220041-872EA 47.6 USD
220041-872 220041-870 220041-871
Dimethyl isophthalate ≥99%
Dimethyl isophthalate
Formula: C₁₀H₁₀O₄
MW: 194.19 g/mol
Boiling Pt: 285 °C (1013 hPa)
Melting Pt: 61 - 63 °C
Density: 1.186 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008433
CAS Number: 1459-93-4
EINECS: 215-951-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR