Benzimidazole

Synonyms: 1H-Benzo[d]imidazole, 1H-Benzimidazole

194123-100G 194123-500G 194123-5G
220041-857EA 56.22 USD
220041-857 220041-858 220041-859
Benzimidazole
Benzimidazole
Formula: C₇H₆N₂
MW: 118.14 g/mol
Boiling Pt: 360 °C (1013 hPa)
Melting Pt: 170 - 173 °C
Density: 1.23 g/cm³ (20 °C)
MDL Number: MFCD00005585
CAS Number: 51-17-2
EINECS: 200-081-4
Merck Index: 13,01082

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR