4-Amino-N,N-dimethylaniline ≥97%

Danger

Synonyms: N,N-Dimethylbenzene-p-diamine, N,N-Dimethyl-1,4-phenylenediamine, N,N-Dimethylbenzene-1,4-diamine, N,N-Dimethyl-p-phenylenediamine

193992-500G 193992-5G 193992-100G
220041-850EA 498.46 USD
220041-850 220041-851 220041-849
4-Amino-N,N-dimethylaniline ≥97%
4-Amino-N,N-dimethylaniline
Formula: C₈H₁₂N₂
MW: 136.2 g/mol
Boiling Pt: 262 °C (1013 hPa)
Melting Pt: 35 °C
Density: 1.09 g/cm³ (20 °C)
Flash Pt: 130 °C
Storage Temperature: Ambient
MDL Number: MFCD00007860
CAS Number: 99-98-9
EINECS: 202-807-5
UN: 2811
ADR: 6.1,II
Merck Index: 13,33280

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR