4-Methylthiazole

193925-100G 193925-25G
220041-842EA 101.23 USD
220041-842 220041-843
4-Methylthiazole
4-Methylthiazole
Formula: C₄H₅NS
MW: 99.16 g/mol
Boiling Pt: 133 °C (1013 hPa)
Density: 1.1187 g/cm³ (20 °C)
MDL Number: MFCD00005340
CAS Number: 693-95-8
EINECS: 211-764-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR