4,4'-Di-tert-butylbiphenyl

Synonyms: 1-tert-butyl-4-(4-tert-butylphenyl)benzene, 4,4'-Di-tert-butyl-1,1'-biphenyl

193801-5G 193801-25G
220041-831EA 64.18 USD
220041-831 220041-830
4,4'-Di-tert-butylbiphenyl
4,4'-Di-tert-butylbiphenyl
Formula: C₂₀H₂₆
MW: 266.43 g/mol
Boiling Pt: 228 °C (1013 hPa)
Melting Pt: 127 °C
Density: 0.9691 g/cm³ (20 °C)
MDL Number: MFCD00008834
CAS Number: 1625-91-8
EINECS: 216-615-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR