2-Chloropyrimidine

Warning

193291-10G 193291-50G
220041-717EA 84 USD
220041-717 220041-718
2-Chloropyrimidine
2-Chloropyrimidine
Formula: C₄H₃ClN₂
MW: 114.53 g/mol
Boiling Pt: 199 °C (1013 hPa)
Melting Pt: 65…66 °C
MDL Number: MFCD00006060
CAS Number: 1722-12-9
EINECS: 217-020-2
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR