α,α,α,α',α',α'-Hexafluoro-3,5-xylidine ≥97%

Warning

Synonyms: 3,5-Di(trifluoromethyl)aniline, 3,5-Bis(trifluoromethyl)aniline

193135-10G 193135-50G
220041-101EA 54.38 USD
220041-101 220041-102
α,α,α,α',α',α'-Hexafluoro-3,5-xylidine ≥97%
α,α,α,α',α',α'-Hexafluoro-3,5-xylidine
Formula: C₈H₅F₆N
MW: 229.12 g/mol
Boiling Pt: 176 °C (1013 hPa)
Melting Pt: 3 °C
Density: 1.4753 g/cm³ (20 °C)
Flash Pt: 83 °C
Storage Temperature: Ambient
MDL Number: MFCD00000394
CAS Number: 328-74-5
EINECS: 206-335-0
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR