(±)-2-Methylbutyric acid ≥98%

Synonyms: (±)-2-Methylbutanoic acid, 2-Methylbutyric acid, 2-Methylbutanoic acid

193070-25G 193070-100G
220041-092EA 42.54 USD
220041-092 220041-091
(±)-2-Methylbutyric acid ≥98%
(±)-2-Methylbutyric acid
Formula: C₅H₁₀O₂
MW: 102.13 g/mol
Boiling Pt: 177 °C (1013 hPa)
Melting Pt: –70 °C
Density: 0.938 g/cm³ (20 °C)
Flash Pt: 80 °C
Storage Temperature: Ambient
MDL Number: MFCD00002669
CAS Number: 116-53-0
EINECS: 204-145-2
UN: 3265
ADR: 8,III

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR