Diisobutylaluminium hydride 25% (w/w) in toluene

Synonyms: Di-i-butylaluminum hydride, DIBAL-H

192724-15KG 192724-7KG 192724-100G 192724-700G
220041-311EA 1820.77 USD
220041-311 220041-313 220041-310 220041-312
Diisobutylaluminium hydride 25% (w/w) in toluene
Diisobutylaluminium hydride
Formula: C₈H₁₉Al
MW: 142.22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00008928
CAS Number: 1191-15-7
EINECS: 214-729-9
UN: 3399
ADR: 4.3,I

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR