trans-2-Methyl-2-butenal ≥96%

Danger

Synonyms: Trans-2-methyl-2-butenal, Tiglic aldehyde, (E)-2-Methyl-2-butenal

192619-5G 192619-25G
220041-088EA 46.2 USD
220041-088 220041-087
trans-2-Methyl-2-butenal ≥96%
trans-2-Methyl-2-butenal
Formula: C₅H₈O
MW: 84.12 g/mol
Boiling Pt: 115…117 °C (1013 hPa)
Density: 0.8688 g/cm³ (20 °C)
MDL Number: MFCD00006977
CAS Number: 497-03-0
EINECS: 207-833-0
UN: 1989
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR