2-(2-Aminophenyl)ethanol ≥97%

Synonyms: 2-(2-Aminophenyl)ethyl alcohol, 2-Aminophenethyl alcohol

192600-25G 192600-5G
220041-711EA 86.05 USD
220041-711 220041-086
2-(2-Aminophenyl)ethanol ≥97%
2-(2-Aminophenyl)ethanol
Formula: C₈H₁₁NO
MW: 137.18 g/mol
Boiling Pt: 308 °C (1013 hPa)
Melting Pt: 89 °C
Density: 1.045 g/cm³ (25 °C)
MDL Number: MFCD00007754
CAS Number: 5339-85-5
EINECS: 226-275-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR