1-Bromotridecane ≥98%

Synonyms: Tridecyl bromide

192422-25G 192422-5G
220041-705EA 565.38 USD
220041-705 220041-706
1-Bromotridecane ≥98%
1-Bromotridecane
Formula: BrH₂C(CH₂)₁₁CH₃
MW: 263.26 g/mol
Boiling Pt: 292 °C (1013 hPa)
Melting Pt: 6 °C
Density: 1.0262 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000227
CAS Number: 765-09-3
EINECS: 212-138-0

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR