4-Bromo-2,6-xylidine ≥98%

Synonyms: 4-Bromo-2,6-dimethylaniline

192376-100G 192376-25G
220041-696EA 268.15 USD
220041-696 220041-697
4-Bromo-2,6-xylidine ≥98%
4-Bromo-2,6-xylidine
Formula: C₈H₁₀BrN
MW: 200.08 g/mol
Boiling Pt: 120 °C (0.2 -rr)
Melting Pt: 50 °C
MDL Number: MFCD00007826
CAS Number: 24596-19-8
EINECS: 246-337-9
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR