(±)-2-Amino-1-propanol ≥98%

Danger

Synonyms: DL-Alaninol, 2-aminopropan-1-ol, DL-2-Amino-1-propanol

192171-25G 192171-5G
220041-682EA 61.49 USD
220041-682 220041-683
(±)-2-Amino-1-propanol ≥98%
(±)-2-Amino-1-propanol
Formula: C₃H₉NO
MW: 75.11 g/mol
Boiling Pt: 173 °C (1013 hPa)
Melting Pt: 8 °C
Density: 0.928 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008085
CAS Number: 6168-72-5
EINECS: 228-207-3
Merck Index: 12,00488

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR