Borane dimethyl sulfide complex 2 mol/L in THF

Synonyms: Borane-methyl sulphide complex

192120-800ML 192120-100ML
220041-680EA 416.08 USD
220041-680 220041-679
Borane dimethyl sulfide complex 2 mol/L in THF
Borane dimethyl sulphide complex
Formula: C₂H₉BS
MW: 75.97 g/mol
MDL Number: MFCD00013189
CAS Number: 13292-87-0
EINECS: 236-313-6
UN: 3399
ADR: 4.3,I

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR