3-Phenoxybenzaldehyde ≥95%

Synonyms: 3-Phenoxybenzenecarbaldehyde, m-Phenoxybenzenecarbaldehyde

191752-25G 191752-5G
220041-266EA 41.57 USD
220041-266 220041-267
3-Phenoxybenzaldehyde ≥95%
3-Phenoxybenzaldehyde
Formula: C₁₃H₁₀O₂
MW: 198.22 g/mol
Boiling Pt: 392 °C (1013 hPa)
Melting Pt: 13 °C
Density: 1.147 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003353
CAS Number: 39515-51-0
EINECS: 254-487-1
UN: 2810
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR