3,5-Dimethoxybenzyl alcohol ≥99%

Synonyms: (3,5-Dimethoxyphenyl)methanol

191655-5G 191655-25G
220041-256EA 848.69 USD
220041-256 220041-255
3,5-Dimethoxybenzyl alcohol ≥99%
3,5-Dimethoxybenzyl alcohol
Formula: C₉H₁₂O₃
MW: 168.19 g/mol
Boiling Pt: 106 °C (0.03 mmHg)
Melting Pt: 48…50 °C
MDL Number: MFCD00004641
CAS Number: 705-76-0
EINECS: 211-888-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR