α-Chloro-3,5-dinitrotoluene ≥97%

Synonyms: 3,5-Dinitrobenzyl chloride

191647-50G 191647-10G
220041-254EA 736.62 USD
220041-254 220041-253
α-Chloro-3,5-dinitrotoluene ≥97%
α-Chloro-3,5-dinitrotoluene
Formula: C₇H₅ClN₂O₄
MW: 216.58 g/mol
Storage Temperature: Ambient
CAS Number: 74367-78-5
EINECS: 277-841-7
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR