4-Aminobenzyl alcohol ≥98%

Synonyms: 4-(Hydroxymethyl)aniline, (4-Aminophenyl)methanol

191558-25G 191558-5G
220040-173EA 498.62 USD
220040-173 220040-174
4-Aminobenzyl alcohol ≥98%
4-Aminobenzyl alcohol
Formula: C₇H₉NO
MW: 123.15 g/mol
Boiling Pt: 169 - 171 °C (11 -rr)
Melting Pt: 75 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00014782
CAS Number: 623-04-1
EINECS: 210-767-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR