3-Aminobenzyl alcohol ≥97%

Synonyms: 3-(Hydroxymethyl)aniline, 3-(Hydroxymethyl)aniline, 3-Aminobenzyl Alcohol, (3-Aminophenyl)methanol

191396-10G 191396-25G
220041-641EA 121.69 USD
220041-641 220041-642
3-Aminobenzyl alcohol ≥97%
3-Aminobenzyl alcohol
Formula: C₇H₉NO
MW: 123.15 g/mol
Boiling Pt: 272 °C (1013 hPa)
Melting Pt: 90 - 93 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00007817
CAS Number: 1877-77-6
EINECS: 217-514-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR