4-Ethylbenzoic acid ≥99%

191280-25G 191280-5G
220041-626EA 92.18 USD
220041-626 220041-627
4-Ethylbenzoic acid ≥99%
4-Ethylbenzoic acid
Formula: C₉H₁₀O₂
MW: 150.18 g/mol
MDL Number: MFCD00002570
CAS Number: 619-64-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR