2-Hydroxy-3-nitropyridine

Synonyms: 3-Nitropyridin-2(1H)-one, 3-Nitro-2-pyridinol

190616-25G 190616-5G
220041-060EA 154.62 USD
220041-060 220041-298
2-Hydroxy-3-nitropyridine
2-Hydroxy-3-nitropyridine
Formula: C₅H₄N₂O₃
MW: 140.1 g/mol
Melting Pt: 224 °C
MDL Number: MFCD00006270
CAS Number: 6332-56-5
EINECS: 228-709-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR