Acrolein dimethyl acetal ≥98%

Synonyms: 3,3-Dimethoxy-1-propene

190543-25ML 190543-5ML
220040-235EA 693.54 USD
220040-235 220040-236
Acrolein dimethyl acetal ≥98%
Acrolein dimethyl acetal
Formula: C₅H₁₀O₂
MW: 102.13 g/mol
Boiling Pt: 89 °C (1013 hPa)
Density: 0.862 g/cm³ (20 °C)
MDL Number: MFCD00008632
CAS Number: 6044-68-4
EINECS: 227-936-4
UN: 3384
ADR: 6.1,I

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR