L-Phenylalaninol ≥98%

Synonyms: (S)-(-)-2-Amino-3-phenyl-1-propanol, (S)-(-)-Phenylalaninol, (S)-2-Amino-3-phenylpropan-1-ol, L(-)-2-Amino-3-phenyl-1-propanol, L(-)-Phenylalaninol, H-Phenylalaninol, L(-)-2-Amino-3-phenylpropan-1-ol, (S)-(-)-2-Amino-3-phenylpropan-1-ol

190438-1G 190438-10G
220039-555EA 43.94 USD
220039-555 220041-059
L-Phenylalaninol ≥98%
L-Phenylalaninol
Formula: C₉H₁₃NO
MW: 151.21 g/mol
Boiling Pt: 278 °C (1013 hPa)
Melting Pt: 91 - 94 °C
MDL Number: MFCD00004732
CAS Number: 3182-95-4
EINECS: 221-674-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR