(±)-1,2,4-Butanetriol, Technical Grade

19040-100ML 19040-25ML
220041-056EA 258.46 USD
220041-056 220041-057
(±)-1,2,4-Butanetriol, Technical Grade
(±)-1,2,4-Butanetriol
Formula: C₄H₁₀O₃
MW: 106.12 g/mol
Boiling Pt: 304 °C (1013 hPa)
Melting Pt: –20 °C
Density: 1.21 g/cm³ (0 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002929
CAS Number: 3068-00-6
EINECS: 221-323-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR