Diisobutylaluminium hydride 1 M in hexane

Synonyms: Di-i-butylaluminum hydride, DIBAL-H

190306-800ML 190306-100ML
220041-053EA 168.92 USD
220041-053 220041-052
Diisobutylaluminium hydride 1 M in hexane
Diisobutylaluminium hydride
Formula: C₈H₁₉Al
MW: 142.22 g/mol
Density: 0.7 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008928
CAS Number: 1191-15-7
EINECS: 214-729-9
UN: 3399
ADR: 4.3,I

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR