3-Phenoxybenzoic acid ≥98%

Warning

190276-5G 190276-25G
220041-049EA 76.03 USD
220041-049 220041-048
3-Phenoxybenzoic acid ≥98%
3-Phenoxybenzoic acid
Formula: C₁₃H₁₀O₃
MW: 214.22 g/mol
Boiling Pt: 365 °C (1013 hPa)
Melting Pt: 149…150 °C
Storage Temperature: Ambient
MDL Number: MFCD00002498
CAS Number: 3739-38-6
EINECS: 223-121-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR