2-Amino-6-nitrobenzothiazole

Synonyms: 6-Nitrobenzothiazol-2-ylamine, 6-Nitro-1,3-benzothiazol-2-amine, 6-Nitrobenzo[d]thiazol-2-amine

190241-100G 190241-25G
220041-045EA 254.15 USD
220041-045 220041-046
2-Amino-6-nitrobenzothiazole
2-Amino-6-nitrobenzothiazole
Formula: C₇H₅N₃O₂S
MW: 195.2 g/mol
Melting Pt: 252 °C
Density: 1.649 g/cm³ (20 °C)
MDL Number: MFCD00005786
CAS Number: 6285-57-0
EINECS: 228-513-7
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR