Pyridinium chlorochromate

Synonyms: PCC

190144-25G 190144-500G 190144-100G
220041-520EA 23.46 USD
220041-520 220041-293 220041-519
Pyridinium chlorochromate
Pyridinium chlorochromate
Formula: C₅H₆ClCrNO₃
MW: 215.56 g/mol
Melting Pt: 205 °C
MDL Number: MFCD00013106
CAS Number: 26299-14-9
EINECS: 247-595-5
UN: 1479
ADR: 5.1,II
Merck Index: 13,08064

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR