2,6-Difluorobenzoic acid ≥98%

Warning

190039-25G 190039-5G
220041-085EA 116.31 USD
220041-085 220041-514
2,6-Difluorobenzoic acid ≥98%
2,6-Difluorobenzoic acid
Formula: C₇H₄F₂O₂
MW: 158.1 g/mol
Boiling Pt: 72 - 77 °C (13 -rr)
Melting Pt: 159 - 160 °C
Storage Temperature: Ambient
MDL Number: MFCD00002411
CAS Number: 385-00-2
EINECS: 206-856-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR