2,3-Dimethoxybenzylamine ≥99%

Synonyms: 1-(2,3-Dimethoxyphenyl)methanamine, (2,3-Dimethoxyphenyl)methanamine

189855-5G 189855-25G
220041-250EA 60 USD
220041-250 220041-249
2,3-Dimethoxybenzylamine ≥99%
2,3-Dimethoxybenzylamine
Formula: C₉H₁₃NO₂
MW: 167.21 g/mol
MDL Number: MFCD00052392
CAS Number: 4393-09-3
EINECS: 224-515-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR