2,3-Dihydroxybenzaldehyde ≥97%

Warning

189839-25G 189839-5G
220041-247EA 205.69 USD
220041-247 220041-248
2,3-Dihydroxybenzaldehyde ≥97%
2,3-Dihydroxybenzaldehyde
Formula: C₇H₆O₃
MW: 138.12 g/mol
Boiling Pt: 235 °C (1013 hPa)
Melting Pt: 103…105 °C
Storage Temperature: Ambient
MDL Number: MFCD00003324
CAS Number: 24677-78-9
EINECS: 246-398-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR